Stampa…

4-Pentynoic acid ≥98.0% (by GC, titration analysis)

Fornitore: TCI EUROPE
Pericolo

P2341-1G P2341-5G
TCIAP2341-1GEA 40 EUR
TCIAP2341-1G TCIAP2341-5G
4-Pentynoic acid ≥98.0% (by GC, titration analysis)
4-Pentynoic acid
Formula: C₅H₆O₂
Peso molecolare: 98,1 g/mol
Punto di ebollizione: 204°C
Temperatura di stoccaggio: Refrigerata
MDL Number: MFCD00004407
CAS Number: 6089-09-4
EINECS: 228-028-0
UN: 3261
ADR: 8,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d6af35d15b652859e3d8ac0b3c9a5f18cbf8f22d&vid=d6af35d15b652859e3d8ac0b3c9a5f18cbf8f22d&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Orange to Green powder to crystal
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 98.0 %
Melting point 55.0 to 59.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR