Stampa…

2-(2-Propynyloxy)benzenecarbaldehyde ≥98.0% (by GC)

Fornitore: TCI EUROPE
Attenzione

Synonyms: 2-(Prop-2-yn-1-yloxy)benzaldehyde, 2-(2-Propynyloxy)benzaldehyde, 2-(Propargyloxy)benzaldehyde

P2338-200MG P2338-1G
TCIAP2338-200MGEA 50 EUR
TCIAP2338-200MG TCIAP2338-1G
2-(2-Propynyloxy)benzenecarbaldehyde ≥98.0% (by GC)
2-(2-Propynyloxy)benzenecarbaldehyde
Formula: C₁₀H₈O₂
Peso molecolare: 160,17 g/mol
Temperatura di stoccaggio: Refrigerata
MDL Number: MFCD02380378
CAS Number: 29978-83-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=7151b65f611c75a7b49927c8b68d0a723f199441&vid=7151b65f611c75a7b49927c8b68d0a723f199441&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Light yellow to Light orange powder to crystal
Purity (GC) min. 98.0 %
Melting point 68.0 to 72.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR