4-(3-Pyridyl)benzaldehyde ≥98.0% (by GC)
Fornitore: TCI
|
Attenzione
|
P2332-200MG
P2332-1G
TCIAP2332-200MGEA
21
EUR
TCIAP2332-200MG
TCIAP2332-1G
4-(3-Pyridyl)benzaldehyde ≥98.0% (by GC)
4-(3-Pyridyl)benzaldehyde
Formula:
C₁₂H₉NO |
MDL Number:
MFCD01631910 CAS Number: 127406-55-7 |
|
Risultati del test di specificazione
Appearance | White to Yellow to Orange powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 52.0 to 56.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...