Stampa…

trans-4-n-Propyl-4'-(3,4,5-trifluorophenyl)bicyclohexyl ≥98.0% (by GC)

Fornitore: TCI EUROPE

Synonyms: trans,trans-4'-Propyl-4-(3,4,5-trifluorophenyl)bicyclohexyl

P2314-5G P2314-25G
TCIAP2314-5GEA 56 EUR
TCIAP2314-5G TCIAP2314-25G
trans-4-n-Propyl-4'-(3,4,5-trifluorophenyl)bicyclohexyl ≥98.0% (by GC)
trans-4-n-Propyl-4'-(3,4,5-trifluorophenyl)bicyclohexyl
Formula: C₂₁H₂₉F₃
Peso molecolare: 338,46 g/mol
MDL Number: MFCD09838998
CAS Number: 131819-23-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=bb220106047a717dbb6fcc2e47a1135ed9a51acf&vid=bb220106047a717dbb6fcc2e47a1135ed9a51acf&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Melting point 64.0 to 68.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR