Stampa…

9H-Pyrido[2,3-b]indole ≥98.0% (by GC, titration analysis)

Fornitore: TCI EUROPE

Synonyms: alpha-Carboline

P2310-200MG P2310-1G
TCIAP2310-200MGEA 50 EUR
TCIAP2310-200MG TCIAP2310-1G
9H-Pyrido[2,3-b]indole ≥98.0% (by GC, titration analysis)
9H-Pyrido[2,3-b]indole
Formula: C₁₁H₈N₂
Peso molecolare: 168,20 g/mol
Punto di ebollizione: 364°C
MDL Number: MFCD00473709
CAS Number: 244-76-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=1717c8bdfa2de33522d76f3dd095c68b5e7fe993&vid=1717c8bdfa2de33522d76f3dd095c68b5e7fe993&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Light yellow to Light orange powder to crystal
Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 98.0 %
Melting point 214.0 to 218.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR