Stampa…

1,4-Phenylenediphosphonic acid ≥98.0% (by HPLC, titration analysis)

Fornitore: TCI EUROPE
Pericolo

Synonyms: 1,4-Benzenediphosphonic acid

P2303-200MG P2303-1G
TCIAP2303-200MGEA 56 EUR
TCIAP2303-200MG TCIAP2303-1G
1,4-Phenylenediphosphonic acid ≥98.0% (by HPLC, titration analysis)
1,4-Phenylenediphosphonic acid
Formula: C₆H₈O₆P₂
Peso molecolare: 238,07 g/mol
MDL Number: MFCD00475582
CAS Number: 880-68-2
UN: 3261
ADR: 8,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=6f2d6b938bcce49afde6345dbd9037053aa00d95&vid=6f2d6b938bcce49afde6345dbd9037053aa00d95&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Neutralization titration) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR