Stampa…

2-Propylpyridine-4-carbothioamide ≥98.0% (by HPLC, titration analysis)

Fornitore: TCI EUROPE
Attenzione

Synonyms: 2-Propyl-4-pyridinecarbothioamide, 2-Propylthioisonicotinamide, 2-Propylpyridine-4-carbothioamide, Prothionamide

P2302-5G P2302-25G
TCIAP2302-5GEA 28 EUR
TCIAP2302-5G TCIAP2302-25G
2-Propylpyridine-4-carbothioamide ≥98.0% (by HPLC, titration analysis)
2-Propylpyridine-4-carbothioamide
Formula: C₉H₁₂N₂S
Peso molecolare: 180,27 g/mol
MDL Number: MFCD00464119
CAS Number: 14222-60-7
EINECS: 238-093-7
Merck Index: 14,07891

Visualizza i codici

Risultati del test di specificazione

Appearance Light yellow to Yellow to Orange powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Nonaqueous Titration) min. 98.0 %
Melting point 141.0 to 145.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR