Stampa…

1,2,3,4,5-Pentaphenyl-1'-(di-tert-butylphosphino)ferrocene CTC-Q-PHOS ≥98.0% (by titrimetric analysis)

Fornitore: TCI EUROPE
Attenzione

Synonyms: CTC-Q-PHOS, 1,2,3,4,5-Pentaphenyl-1'-(di-tert-butylphosphino)ferrocene, QPhos, 1,2,3,4,5-Pentaphenyl-1'-(di-t-butylphosphino)ferrocene

P2297-100MG P2297-500MG
TCIAP2297-100MGEA 161 EUR
TCIAP2297-100MG TCIAP2297-500MG
1,2,3,4,5-Pentaphenyl-1'-(di-tert-butylphosphino)ferrocene CTC-Q-PHOS ≥98.0% (by titrimetric analysis)
1,2,3,4,5-Pentaphenyl-1'-(di-tert-butylphosphino)ferrocene CTC-Q-PHOS
Formula: C₄₈H₄₇FeP
Peso molecolare: 710,706 g/mol
MDL Number: MFCD03788940
CAS Number: 312959-24-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d89ed3db8cfe06deb724568e0d46c17607afa80e&vid=d89ed3db8cfe06deb724568e0d46c17607afa80e&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance Orange to Brown powder to crystal
Purity (Chelometric Titration) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR