Stampa…

2-Phenylbenzoxazole ≥98.0% (by GC)

Fornitore: TCI
Attenzione

P2264-1G P2264-5G
TCIAP2264-1GEA 71 EUR
TCIAP2264-1G TCIAP2264-5G
2-Phenylbenzoxazole ≥98.0% (by GC)
2-Phenylbenzoxazole
Formula: C₁₃H₉NO
MDL Number: MFCD00012183
CAS Number: 833-50-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=0897c9c625cf4ebc4bcb00bd2af065ffaa51aef0&vid=0897c9c625cf4ebc4bcb00bd2af065ffaa51aef0&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Light yellow to Dark green powder to crystal
Purity (GC) min. 98.0 %
Melting point 101.0 to 105.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR