1H-Pyrazole-3-carbaldehyde ≥98.0% (by GC)
Fornitore: TCI EUROPE
|
Attenzione
|
Synonyms:
3-Formylpyrazole, Pyrazole-3-carboxaldehyde, 3-Formyl-1H-pyrazole
P2261-200MG
P2261-1G
TCIAP2261-200MGEA
48
EUR
TCIAP2261-200MG
TCIAP2261-1G
1H-Pyrazole-3-carbaldehyde ≥98.0% (by GC)
1H-Pyrazole-3-carbaldehyde
Formula:
C₄H₄N₂O Peso molecolare: 96,09 g/mol |
MDL Number:
MFCD00129925 CAS Number: 3920-50-1 |
Risultati del test di specificazione
Appearance | White to Orange to Green powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 149.0 to 153.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...