Stampa…

2-Propynyl [3-(Triethoxysilyl)propyl]carbamate ≥90.0% (by GC)

Fornitore: TCI EUROPE
Attenzione

Synonyms: [3-(Triethoxysilyl)propyl]carbamic acid Propargyl ester, [3-(Triethoxysilyl)propyl]carbamic acid 2-Propynyl ester, Propargyl [3-(Triethoxysilyl)propyl]carbamate, Prop-2-yn-1-yl (3-(triethoxysilyl)propyl)carbamate

P2258-1G P2258-5G
TCIAP2258-1GEA 114 EUR
TCIAP2258-1G TCIAP2258-5G
2-Propynyl [3-(Triethoxysilyl)propyl]carbamate ≥90.0% (by GC)
2-Propynyl [3-(Triethoxysilyl)propyl]carbamate
Formula: C₁₃H₂₅NO₅Si
Peso molecolare: 303,43 g/mol
MDL Number: MFCD01320971
CAS Number: 870987-68-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c77d0653f0a6926674d3c68a82732a91be345119&vid=c77d0653f0a6926674d3c68a82732a91be345119&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance Light orange to Yellow to Green clear liquid
Purity (GC) min. 90.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR