Stampa…

Phenylsuccinic anhydride ≥98.0% (by GC, titration analysis)

Fornitore: TCI
Attenzione

P2257-5G P2257-25G
TCIAP2257-5GEA 92 EUR
TCIAP2257-5G TCIAP2257-25G
Phenylsuccinic anhydride ≥98.0% (by GC, titration analysis)
Phenylsuccinic anhydride
Formula: C₁₀H₈O₃
Punto di ebollizione: 195°C
MDL Number: MFCD00192395
CAS Number: 1131-15-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e93617aa5980588c7e36f51e6d6e4b898d8f820d&vid=e93617aa5980588c7e36f51e6d6e4b898d8f820d&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Orange to Green powder to crystal
Purity (GC) min. 98.0 %
Purity (Methanolysis Method) min. 98.0 %
Melting point 52.0 to 56.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR