Stampa…

2,5,8,11,14-Pentaoxaheptadec-16-yne ≥98.0% (by GC)

Fornitore: TCI EUROPE

Synonyms: mPEG4-Alkyne

P2249-25MG P2249-100MG
TCIAP2249-25MGEA 34 EUR
TCIAP2249-25MG TCIAP2249-100MG
2,5,8,11,14-Pentaoxaheptadec-16-yne ≥98.0% (by GC)
2,5,8,11,14-Pentaoxaheptadec-16-yne
Formula: C₁₂H₂₂O₅
Peso molecolare: 246,30 g/mol
Temperatura di stoccaggio: Refrigerata
CAS Number: 1101668-39-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=44e126e6e2b58d83e29e5cb598343f67725432ee&vid=44e126e6e2b58d83e29e5cb598343f67725432ee&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance Colorless to Light yellow clear liquid
Purity (GC) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR