1-(1,3-Thiazol-2-yl)propan-1-one ≥98.0% (by GC)
Fornitore: TCI EUROPE
Synonyms:
2-Propionylthiazole, 1-(Thiazol-2-yl)propan-1-one
P2247-5G
P2247-25G
TCIAP2247-5GEA
55
EUR
TCIAP2247-5G
TCIAP2247-25G
1-(1,3-Thiazol-2-yl)propan-1-one ≥98.0% (by GC)
1-(1,3-Thiazol-2-yl)propan-1-one
Formula:
C₆H₇NOS Peso molecolare: 141,19 g/mol Punto di ebollizione: 107°C |
MDL Number:
MFCD01681404 CAS Number: 43039-98-1 |
Risultati del test di specificazione
Appearance | Colorless to Light yellow to Light orange clear liquid |
Purity (GC) | min. 98.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...