Polyethylene glycol dimethylether, M.W. ∼ 240
Fornitore: TCI
Formula:
H₃C(OCH₂CH₂)nOCH₃ Flash Pt: 156°C |
MDL Number:
MFCD00147972 CAS Number: 24991-55-7 |
|
Risultati del test di specificazione
Appearance | Colorless to Almost colorless clear liquid |
Average molecular weight | 230 to 250 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...