Stampa…

2-(1-Pyrazolyl)pyridine ≥98.0% (by GC)

Fornitore: TCI EUROPE
Attenzione

Synonyms: 2-(1H-Pyrazol-1-yl)pyridine

P2239-1G P2239-5G
TCIAP2239-1GEA 61 EUR
TCIAP2239-1G TCIAP2239-5G
2-(1-Pyrazolyl)pyridine ≥98.0% (by GC)
2-(1-Pyrazolyl)pyridine
Formula: C₈H₇N₃
Peso molecolare: 145,17 g/mol
MDL Number: MFCD09033534
CAS Number: 25700-11-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=87a6e27aa92d99f8bf2eaeb7b8f1793eb4f32918&vid=87a6e27aa92d99f8bf2eaeb7b8f1793eb4f32918&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to lump
Purity (GC) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR