Palladio 5% on alumina
Fornitore: TCI EUROPE
|
Pericolo
|
P2238-5G
P2238-25G
TCIAP2238-5GEA
199
EUR
TCIAP2238-5G
TCIAP2238-25G
Palladio 5% on alumina
Palladium
Formula:
Pd Peso molecolare: 106,420 g/mol |
MDL Number:
MFCD03613602 CAS Number: 7440-05-3 EINECS: 231-115-6 Merck Index: 14,06989 |
Risultati del test di specificazione
Appearance | Black powder to crystal |
Palladium | 4.90 to 5.10 % |
Water | max. 2.00 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...