2-[2-(2-Propynyloxy)ethoxy]ethylamine ≥98.0% (by GC, titration analysis)
Fornitore: TCI EUROPE
|
Pericolo
|
Synonyms:
Propargyl-PEG2-amine, 2-[2-(Propargyloxy)ethoxy]ethylamine
P2225-1G
P2225-5G
TCIAP2225-1GEA
76
EUR
TCIAP2225-1G
TCIAP2225-5G
2-[2-(2-Propynyloxy)ethoxy]ethylamine ≥98.0% (by GC, titration analysis)
2-[2-(2-Propynyloxy)ethoxy]ethylamine
Formula:
C₇H₁₃NO₂ Peso molecolare: 143,19 g/mol Temperatura di stoccaggio: Refrigerata |
MDL Number:
MFCD19160595 CAS Number: 944561-44-8 UN: 2735 ADR: 8,II |
Risultati del test di specificazione
Appearance | Colorless to Almost colorless clear liquid |
Purity (GC) | min. 98.0 % |
Purity (Neutralization titration) | min. 98.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...