Stampa…

2-(1H-Pyrazol-3-yl)pyridine ≥98.0% (by GC, titration analysis)

Fornitore: TCI EUROPE
Attenzione

P2211-1G P2211-5G
TCIAP2211-1GEA 53 EUR
TCIAP2211-1G TCIAP2211-5G
2-(1H-Pyrazol-3-yl)pyridine ≥98.0% (by GC, titration analysis)
2-(1H-Pyrazol-3-yl)pyridine
Formula: C₈H₇N₃
Peso molecolare: 145,16 g/mol
MDL Number: MFCD00115151
CAS Number: 75415-03-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=10b8dd9668b42d573afcc240164a28fa5bd0f552&vid=10b8dd9668b42d573afcc240164a28fa5bd0f552&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Light yellow powder to crystal
Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 98.0 %
Melting point 122.0 to 126.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR