Stampa…

Pentafluorophenyl Acetate ≥98.0% (by GC)

Fornitore: TCI
Attenzione

Synonyms: Acetic acid Pentafluorophenyl ester

P2192-1G P2192-5G
TCIAP2192-1GEA 53 EUR
TCIAP2192-1G TCIAP2192-5G
Pentafluorophenyl Acetate ≥98.0% (by GC)
Pentafluorophenyl Acetate
Formula: C₈H₃F₅O₂
Peso molecolare: 226.10 g/mol
Punto di ebollizione: 60°C
MDL Number: MFCD00015552
CAS Number: 19220-93-0
EINECS: 242-891-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c683f5722ea762ca3e04a97ed1e6d288d366f5f0&vid=c683f5722ea762ca3e04a97ed1e6d288d366f5f0&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to lump
Purity (GC) min. 98.0 %
Melting point 30.0 to 34.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR