N-(2-Propynyl)succinimide ≥98.0% (by GC)
Fornitore: TCI EUROPE
|
Attenzione
|
Synonyms:
1-(Prop-2-yn-1-yl)pyrrolidine-2,5-dione, N-Propargylsuccinimide
P2191-200MG
P2191-1G
TCIAP2191-200MGEA
92
EUR
TCIAP2191-200MG
TCIAP2191-1G
N-(2-Propynyl)succinimide ≥98.0% (by GC)
N-(2-Propynyl)succinimide
Formula:
C₇H₇NO₂ Peso molecolare: 137,14 g/mol Punto di ebollizione: 237°C |
MDL Number:
MFCD06740819 CAS Number: 10478-33-8 |
Risultati del test di specificazione
Appearance | White to Light yellow powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 51.0 to 55.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...