Stampa…

2-(2-Propynyloxy)naphthalene ≥98.0% (by GC)

Fornitore: TCI

Synonyms: 2-(Prop-2-yn-1-yloxy)naphthalene, 2-(Propargyloxy)naphthalene

P2190-200MG P2190-1G
TCIAP2190-200MGEA 73 EUR
TCIAP2190-200MG TCIAP2190-1G
2-(2-Propynyloxy)naphthalene ≥98.0% (by GC)
2-(2-Propynyloxy)naphthalene
Formula: C₁₃H₁₀O
Punto di ebollizione: 83°C
MDL Number: MFCD04275627
CAS Number: 20009-28-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d83fc6fceb6cd33bb7bc4e314e7adb8bba552b90&vid=d83fc6fceb6cd33bb7bc4e314e7adb8bba552b90&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Melting point 58.0 to 62.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR