Stampa…

1-Phenylcyclohexanecarboxylic acid ≥95.0% (by GC, titration analysis)

Fornitore: TCI
Attenzione

Synonyms: 1-Phenyl-1-cyclohexanecarboxylic acid

P2180-1G P2180-5G
TCIAP2180-1GEA 33 EUR
TCIAP2180-1G TCIAP2180-5G
1-Phenylcyclohexanecarboxylic acid ≥95.0% (by GC, titration analysis)
1-Phenylcyclohexanecarboxylic acid
Formula: C₁₃H₁₆O₂
Peso molecolare: 204.27 g/mol
MDL Number: MFCD00037152
CAS Number: 1135-67-7
EINECS: 214-495-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=33c8608906944dda202ac7cd173018ce1f207601&vid=33c8608906944dda202ac7cd173018ce1f207601&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Light yellow powder to crystal
Purity (GC) min. 95.0 %
Purity (Neutralization titration) min. 95.0 %
Melting point 121.0 to 125.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR