Stampa…

(1,10-Phenanthroline)(trifluoromethyl)(triphenylphosphine)copper(I) ≥95.0% (by titrimetric analysis)

Fornitore: TCI EUROPE
Attenzione

P2172-1G P2172-5G
TCIAP2172-1GEA 240 EUR
TCIAP2172-1G TCIAP2172-5G
(1,10-Phenanthroline)(trifluoromethyl)(triphenylphosphine)copper(I) ≥95.0% (by titrimetric analysis)
(1,10-Phenanthroline)(trifluoromethyl)(triphenylphosphine)copper(I)
Formula: C₃₁H₂₃CuF₃N₂P
Peso molecolare: 575,05 g/mol
MDL Number: MFCD28386111
CAS Number: 1334890-93-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8c109c7512b13cf4de3c5dfbeab046c1a4cb2c55&vid=8c109c7512b13cf4de3c5dfbeab046c1a4cb2c55&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance Light yellow to Brown powder to crystal
Purity (Chelometric Titration) min. 95.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR