2-(2-Propynyloxy)ethylamine ≥98.0% (by titrimetric analysis)
Fornitore: TCI EUROPE
|
Pericolo
|
Synonyms:
Propargyl-PEG1-amine, Propargyl-PEG1-NH2
P2166-1G
P2166-5G
TCIAP2166-1GEA
366
EUR
TCIAP2166-1G
TCIAP2166-5G
2-(2-Propynyloxy)ethylamine ≥98.0% (by titrimetric analysis)
2-(2-Propynyloxy)ethylamine
Formula:
C₅H₉NO Peso molecolare: 99,13 g/mol Temperatura di stoccaggio: Refrigerata |
MDL Number:
MFCD16713446 CAS Number: 122116-12-5 UN: 2734 ADR: 8,II |
Risultati del test di specificazione
Appearance | Colorless to Light orange to Yellow clear liquid |
Purity (GC) | min. 96.0 % |
Purity (Nonaqueous Titration) | min. 98.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...