Stampa…

Penciclovir ≥98.0% (by HPLC, titration analysis)

Fornitore: TCI EUROPE

Synonyms: 2-Amino-1,9-dihydro-9-[4-hydroxy-3-(hydroxymethyl)butyl]-6H-purin-6-one, 2-Amino-9-[4-hydroxy-3-(hydroxymethyl)butyl]-1H-purin-6-one, 9-[4-Hydroxy-3-(hydroxymethyl)butyl]guanine, BRL 39123, 2-Amino-9-(4-hydroxy-3-(hydroxymethyl)butyl)-1H-purin-6(9H)-one

P2164-200MG P2164-1G
TCIAP2164-200MGEA 58 EUR
TCIAP2164-200MG TCIAP2164-1G
Penciclovir ≥98.0% (by HPLC, titration analysis)
Penciclovir
Formula: C₁₀H₁₅N₅O₃
Peso molecolare: 253,26 g/mol
Temperatura di stoccaggio: Congelamento
MDL Number: MFCD15142866
CAS Number: 39809-25-1
Merck Index: 14,07083
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=35a8cd9e9544fd2327637f8d98e4cc9c7036edce&vid=35a8cd9e9544fd2327637f8d98e4cc9c7036edce&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Nonaqueous Titration) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR