Stampa…

4-(trans-4-Propylcyclohexyl)phenylboronic acid (contains varying amounts of Anhydride)

Fornitore: TCI EUROPE
Attenzione

Synonyms: (4-(trans-4-propylcyclohexyl)phenyl)boronic acid

P2162-1G P2162-5G
TCIAP2162-1GEA 19 EUR
TCIAP2162-1G TCIAP2162-5G
4-(trans-4-Propylcyclohexyl)phenylboronic acid (contains varying amounts of Anhydride)
4-(trans-4-Propylcyclohexyl)phenylboronic acid
Formula: C₁₅H₂₃BO₂
Peso molecolare: 246,1529 g/mol
MDL Number: MFCD06658197
CAS Number: 146862-02-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b384ae047ffeafa897669aabd413aea4277dab6d&vid=b384ae047ffeafa897669aabd413aea4277dab6d&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (Neutralization titration) 97.0 to 108.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR