Stampa…

4-(1-Phenyl-1H-benzimidazol-2-yl)phenylboronic acid (contains varying amounts of Anhydride)

Fornitore: TCI EUROPE
Attenzione

Synonyms: (4-(1-Phenyl-1H-benzo[d]imidazol-2-yl)phenyl)boronic acid, 4-(1-Phenyl-1H-benzimidazol-2-yl)benzeneboronic Acid

P2158-1G P2158-5G
TCIAP2158-1GEA 46 EUR
TCIAP2158-1G TCIAP2158-5G
4-(1-Phenyl-1H-benzimidazol-2-yl)phenylboronic acid (contains varying amounts of Anhydride)
4-(1-Phenyl-1H-benzimidazol-2-yl)phenylboronic acid
Formula: C₁₉H₁₅BN₂O₂
Peso molecolare: 314,15 g/mol
MDL Number: MFCD11045057
CAS Number: 952514-79-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=41bb3539c18b6eb6bd5a2eadf57cf98bad440257&vid=41bb3539c18b6eb6bd5a2eadf57cf98bad440257&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Light yellow to Light orange powder to crystal
Purity (Neutralization titration) 95.0 to 107.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR