Stampa…

Potassium trifluoroacetate ≥98.0% (by titrimetric analysis)

Fornitore: TCI
Pericolo

Synonyms: Trifluoroacetic acid potassium salt, Potassium 2,2,2-trifluoroacetate

P2114-25G P2114-100G
TCIAP2114-25GEA 32 EUR
TCIAP2114-25G TCIAP2114-100G
Potassium trifluoroacetate ≥98.0% (by titrimetric analysis)
Potassium trifluoroacetate
Formula: C₂F₃KO₂
MDL Number: MFCD00013215
CAS Number: 2923-16-2
EINECS: 220-877-5
UN: 2811
ADR: 6.1,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=34cfb94ade4999f0bdd31210b94555446049b3bb&vid=c9334f8fc0882f53b243cbff9ea144536813ba53&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Light yellow powder to crystal
Purity (Ion exchange titration) min. 98.0 %
Melting point 140.0 to 144.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR