trans-4-(4-n-Propylcyclohexyl)cyclohexanone ≥98.0% (by GC)
Fornitore: TCI EUROPE
Synonyms:
4-(trans-4-Propylcyclohexyl)cyclohexanone
P2108-1G
P2108-5G
TCIAP2108-1GEA
18
EUR
TCIAP2108-1G
TCIAP2108-5G
trans-4-(4-n-Propylcyclohexyl)cyclohexanone ≥98.0% (by GC)
trans-4-(4-n-Propylcyclohexyl)cyclohexanone
Formula:
C₁₅H₂₆O Peso molecolare: 222,372 g/mol |
MDL Number:
MFCD04038797 CAS Number: 82832-73-3 |
Risultati del test di specificazione
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 98.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...