Stampa…

1,10-Fenantrolina ≥98.0% (by HPLC, titration analysis)

Fornitore: TCI
Pericolo

Synonyms: o-Phenanthroline

P2099-25G P2099-5G
TCIAP2099-25GEA 76 EUR
TCIAP2099-25G TCIAP2099-5G
1,10-Fenantrolina ≥98.0% (by HPLC, titration analysis)
1,10-Fenantrolina
Formula: C₁₂H₈N₂
MDL Number: MFCD00011678
CAS Number: 66-71-7
EINECS: 200-629-2
UN: 2811
ADR: 6.1,III
Merck Index: 14,07214
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5092a8ec81301ed6ed30f75429f9f27784db47a5&vid=106c4325bb7c310de902d3118cf4b31bcee3dc92&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Orange to Green powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Nonaqueous Titration) min. 98.0 %
Melting point 118.0 to 122.0 deg-C
Water max. 1.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR