Phenylmaleic anhydride ≥98.0% (by GC, titration analysis)
Fornitore: TCI
|
Attenzione
|
Synonyms:
3-Phenylfuran-2,5-dione, 3-Phenyl-2,5-furandione
Formula:
C₁₀H₆O₃ Peso molecolare: 174.16 g/mol |
MDL Number:
MFCD00015472 CAS Number: 36122-35-7 EINECS: 252-881-8 |
|
Risultati del test di specificazione
Appearance | White to Light yellow powder to crystal |
Purity (GC) | min. 98.0 % |
Purity (Methanolysis Method) | min. 98.0 % |
Melting point | 120.0 to 124.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...