Stampa…

Pentaerythritol tetraacrylate ≥80.0% (by GC) stabilizzato

Fornitore: TCI EUROPE
Attenzione


Stabilization: Stabilised with 4-methoxyphenol (MEHQ)

P2084-25G P2084-500G
TCIAP2084-25GEA 17 EUR
TCIAP2084-25G TCIAP2084-500G
Pentaerythritol tetraacrylate ≥80.0% (by GC) stabilizzato
Pentaerythritol tetraacrylate
Formula: C₁₇H₂₀O₈
Peso molecolare: 352,34 g/mol
MDL Number: MFCD00015334
CAS Number: 4986-89-4
EINECS: 225-644-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=50c42887097a03e99d7241ab8af4ebd2c3adfc37&vid=50c42887097a03e99d7241ab8af4ebd2c3adfc37&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White or Colorless to Light yellow powder to lump to clear liquid
Purity (GC) min. 80.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR