Stampa…
Prulifloxacin ≥98.0% (by HPLC, titration analysis)
Fornitore: TCI EUROPE
Synonyms: 6-Fluoro-1-methyl-7-(4-((5-methyl-2-oxo-1,3-dioxol-4-yl)methyl)piperazin-1-yl)-4-oxo-1,4-dihydro-[1,3]thiazeto[3,2-a]quinoline-3-carboxylic acid
P2058-1G P2058-5G
TCIAP2058-1GEA 82 EUR
TCIAP2058-1G TCIAP2058-5G
Prulifloxacin ≥98.0% (by HPLC, titration analysis)
Prulifloxacin
Formula: C₂₁H₂₀FN₃O₆S
Peso molecolare: 461,46 g/mol
Punto di fusione: 220°C
Temperatura di stoccaggio: Congelamento
MDL Number: MFCD00864847
CAS Number: 123447-62-1
Merck Index: 14,07908
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=1be4ad54fef9e174be16abd59a3fccc65e81785c&vid=1be4ad54fef9e174be16abd59a3fccc65e81785c&prid=web&clid=SAPDAM
Visualizza i codici
Appearance Light orange to Yellow to Green powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Nonaqueous Titration) min. 98.0 %