2-Phenylquinoline ≥98.0% (by GC, titration analysis)
Fornitore: TCI EUROPE
|
Attenzione
|
P2057-1G
P2057-5G
TCIAP2057-1GEA
54
EUR
TCIAP2057-1G
TCIAP2057-5G
2-Phenylquinoline ≥98.0% (by GC, titration analysis)
2-Phenylquinoline
Formula:
C₁₅H₁₁N Peso molecolare: 205,26 g/mol Punto di ebollizione: 363°C |
MDL Number:
MFCD00011568 CAS Number: 612-96-4 EINECS: 210-326-7 |
Risultati del test di specificazione
Appearance | White to Orange to Green powder to crystal |
Purity (GC) | min. 98.0 % |
Purity (Nonaqueous Titration) | min. 98.0 % |
Melting point | 81.0 to 85.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...