Stampa…

Pentoxifylline ≥98.0% (by HPLC, titration analysis)

Fornitore: TCI EUROPE
Attenzione

Synonyms: 3,7-Dimethyl-1-(5-oxohexyl)xanthine, 1-(5-Oxohexyl)theobromine, 3,7-Dimethyl-1-(5-oxohexyl)-1H-purine-2,6(3H,7H)-dione, 3,7-Dihydro-3,7-dimethyl-1-(5-oxohexyl)-1H-purine-2,6-dione

P2050-5G P2050-25G
TCIAP2050-5GEA 27 EUR
TCIAP2050-5G TCIAP2050-25G
Pentoxifylline ≥98.0% (by HPLC, titration analysis)
Pentoxifylline
Formula: C₁₃H₁₈N₄O₃
Peso molecolare: 278,31 g/mol
MDL Number: MFCD00063379
CAS Number: 6493-05-6
EINECS: 229-374-5
Merck Index: 14,07136
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=986dda3c0d39b5b03898793cbf180abe3177d912&vid=986dda3c0d39b5b03898793cbf180abe3177d912&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Nonaqueous Titration) min. 98.0 %
Melting point 103.0 to 107.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR