Stampa…

Pifithrin-mu ≥98.0% (by HPLC, total nitrogen)

Fornitore: TCI EUROPE
Attenzione

Synonyms: 2-Phenylethynesulphonamide, Pifithrin-µ, Pifithrin-μ, 2-Phenylethyne-1-sulphonamide

P2048-10MG P2048-100MG
TCIAP2048-10MGEA 30 EUR
TCIAP2048-10MG TCIAP2048-100MG
Pifithrin-mu ≥98.0% (by HPLC, total nitrogen)
Pifithrin-mu
Formula: C₈H₇NO₂S
Peso molecolare: 181,21 g/mol
Temperatura di stoccaggio: Refrigerata
MDL Number: MFCD00181531
CAS Number: 64984-31-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=356fa0f42defdedc2f20c7e41423846764abce26&vid=356fa0f42defdedc2f20c7e41423846764abce26&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (with Total Nitrogen) min. 98.0 %
Melting point 135.0 to 139.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR