Stampa…

Prilocaine hydrochloride ≥98.0% (by HPLC)

Fornitore: TCI EUROPE

Synonyms: Prilocaine HCl, 2-(Propylamino)-N-(o-tolyl)propanamide hydrochloride, 2-(Propylamino)-N-(o-tolyl)propionamide Hydrochloride

P2038-1G P2038-5G
TCIAP2038-1GEA 50 EUR
TCIAP2038-1G TCIAP2038-5G
Prilocaine hydrochloride ≥98.0% (by HPLC)
Prilocaine hydrochloride
Formula: C₁₃H₂₀N₂O
Peso molecolare: 256,77 g/mol
MDL Number: MFCD00079279
CAS Number: 1786-81-8
EINECS: 217-244-0
Merck Index: 14,07743
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a707f1fe0bf9c75341b933c8c37f9516dbea2604&vid=a707f1fe0bf9c75341b933c8c37f9516dbea2604&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Nonaqueous Titration) min. 97.0 %
Melting point 164.0 to 168.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR