Stampa…

Pyrimethamine ≥98.0% (by HPLC, titration analysis)

Fornitore: TCI EUROPE
Attenzione

Synonyms: 5-(4-Chlorophenyl)-6-ethylpyrimidine-2,4-diamine, 5-(4-Chlorophenyl)-6-ethyl-2,4-pyrimidinediamine

P2037-1G P2037-5G
TCIAP2037-1GEA 20 EUR
TCIAP2037-1G TCIAP2037-5G
Pyrimethamine ≥98.0% (by HPLC, titration analysis)
Pyrimethamine
Formula: C₁₂H₁₃ClN₄
Peso molecolare: 248,71 g/mol
MDL Number: MFCD00057350
CAS Number: 58-14-0
EINECS: 200-364-2
UN: 3077
ADR: 9,III
Merck Index: 14,07985
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f89c7778518767a98eacc0b76ad47eb1faaeb68a&vid=f89c7778518767a98eacc0b76ad47eb1faaeb68a&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Nonaqueous Titration) min. 98.0 %
Melting point 239.0 to 243.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR