Stampa…

5-Phenylbenzoxazole-2-thiol ≥98.0% (by GC, titration analysis)

Fornitore: TCI
Attenzione

Synonyms: 2-Mercapto-5-phenylbenzoxazole, 5-Phenylbenzoxazoline-2(3H)-thione

P2011-1G P2011-5G
TCIAP2011-1GEA 46 EUR
TCIAP2011-1G TCIAP2011-5G
5-Phenylbenzoxazole-2-thiol ≥98.0% (by GC, titration analysis)
5-Phenylbenzoxazole-2-thiol
Formula: C₁₃H₉NOS
MDL Number: MFCD00022847
CAS Number: 17371-99-2

Visualizza i codici

Risultati del test di specificazione

Appearance White to Light yellow to Dark green powder to crystal
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 98.0 %
Melting point 211.0 to 215.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR