Stampa…

2-Phenylethyl cinnamate ≥99.0% (by GC)

Fornitore: TCI EUROPE

Synonyms: Phenethyl cinnamate, Cinnamic Acid 2-Phenylethyl Ester, Phenethyl Cinnamate, Cinnamic Acid Phenethyl Ester

P2007-25G P2007-500G
TCIAP2007-25GEA 16 EUR
TCIAP2007-25G TCIAP2007-500G
2-Phenylethyl cinnamate ≥99.0% (by GC)
2-Phenylethyl cinnamate
Formula: C₁₇H₁₆O₂
Peso molecolare: 252,31 g/mol
MDL Number: MFCD00022050
CAS Number: 103-53-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e494a5b0536158849d997a08c6040fe6ce7f89d3&vid=e494a5b0536158849d997a08c6040fe6ce7f89d3&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Light yellow powder to crystal
Purity (GC) min. 99.0 %
Melting point 55.0 to 59.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR