Stampa…

4-(4-Pyridyl)benzaldehyde ≥98.0% (by GC)

Fornitore: TCI EUROPE
Attenzione

Synonyms: 4-(Pyridin-4-yl)benzaldehyde

P1983-200MG P1983-1G
TCIAP1983-200MGEA 91 EUR
TCIAP1983-200MG TCIAP1983-1G
4-(4-Pyridyl)benzaldehyde ≥98.0% (by GC)
4-(4-Pyridyl)benzaldehyde
Formula: C₁₂H₉NO
Peso molecolare: 183,21 g/mol
MDL Number: MFCD02684110
CAS Number: 99163-12-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=78d9b3deeb6d3ec1d8f893d9ee75d3b7b7ff99a8&vid=78d9b3deeb6d3ec1d8f893d9ee75d3b7b7ff99a8&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Orange to Green powder to crystal
Purity (GC) min. 98.0 %
Melting point 87.0 to 91.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR