Parthenolide ≥97.0% (by HPLC)
Fornitore: TCI EUROPE
Synonyms:
4,5alpha-Epoxy-6beta-hydroxygermacra-1(10),11(13)-dien-12-oic Acid gamma-Lactone, (3aS,9aR,10aR,10bS,E)-6,9a-Dimethyl-3-methylene-3a,4,5,8,9,9a,10a,10b-octahydrooxireno[2',3':9,10]cyclodeca[1,2-b]furan-2(3H)-one
P1982-100MG
P1982-25MG
TCIAP1982-100MGEA
145
EUR
TCIAP1982-100MG
TCIAP1982-25MG
Parthenolide ≥97.0% (by HPLC)
Parthenolide
Formula:
C₁₅H₂₀O₃ Peso molecolare: 248,32 g/mol Temperatura di stoccaggio: Refrigerata |
MDL Number:
MFCD00134592 CAS Number: 20554-84-1 Merck Index: 14,07048 |
Risultati del test di specificazione
Appearance | White to Light yellow powder to crystal |
Purity (HPLC) | min. 97.0 area% |
Specific rotation [a]20/D | -81.0 to -86.0 deg (C=1, CHCl3) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...