1-Phenyl-3-(2,4,6-trimethylphenyl)-2-pyrazoline ≥98.0% (by GC)
Fornitore: TCI EUROPE
Synonyms:
3-Mesityl-1-phenyl-2-pyrazoline, 3-Mesityl-1-phenyl-4,5-dihydro-1H-pyrazole
P1951-1G
P1951-5G
TCIAP1951-1GEA
100
EUR
TCIAP1951-1G
TCIAP1951-5G
1-Phenyl-3-(2,4,6-trimethylphenyl)-2-pyrazoline ≥98.0% (by GC)
1-Phenyl-3-(2,4,6-trimethylphenyl)-2-pyrazoline
Formula:
C₁₈H₂₀N₂ Peso molecolare: 264,37 g/mol |
MDL Number:
MFCD00153039 CAS Number: 60078-97-9 EINECS: 262-048-0 |
Risultati del test di specificazione
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 95.0 to 99.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...