2-Phenylpyrimidine ≥98.0% (by GC)
Fornitore: TCI EUROPE
|
Attenzione
|
P1945-200MG
P1945-1G
TCIAP1945-200MGEA
117
EUR
TCIAP1945-200MG
TCIAP1945-1G
2-Phenylpyrimidine ≥98.0% (by GC)
2-Phenylpyrimidine
Formula:
C₁₀H₈N₂ Peso molecolare: 156,19 g/mol Punto di ebollizione: 100°C |
MDL Number:
MFCD04038761 CAS Number: 7431-45-0 |
Risultati del test di specificazione
Appearance | White to Almost white powder to lump |
Purity (GC) | min. 98.0 % |
Purity (Nonaqueous Titration) | min. 97.0 % |
Melting point | 37.0 to 41.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...