Stampa…

4-(1-Pyrrolidinyl)piperidine ≥98.0% (by GC, titration analysis)

Fornitore: TCI
Attenzione

P1932-25G P1932-5G
TCIAP1932-25GEA 318 EUR
TCIAP1932-25G TCIAP1932-5G
4-(1-Pyrrolidinyl)piperidine ≥98.0% (by GC, titration analysis)
4-(1-Pyrrolidinyl)piperidine
Formula: C₉H₁₈N₂
MDL Number: MFCD00038011
CAS Number: 5004-07-9
EINECS: 225-676-6

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 98.0 %
Melting point 58.0 to 62.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR