Pentafluorophenylboronic acid (contains varying amounts of Anhydride)
Fornitore: TCI
|
Attenzione
|
Formula:
C₆H₂BF₅O₂ Punto di fusione: 290°C |
MDL Number:
MFCD01074663 CAS Number: 1582-24-7 |
|
Risultati del test di specificazione
Appearance | White to Almost white powder to crystal |
Purity (Neutralization titration) | 97.0 to 110.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...