Stampa…

Paliperidone ≥97.0% (by titrimetric analysis)

Fornitore: TCI EUROPE
Pericolo

Synonyms: 3-[2-[4-(6-Fluoro-1,2-benzisoxazol-3-yl)-1-piperidinyl]ethyl]-6,7,8,9-tetrahydro-9-hydroxy-2-methyl-4H-pyrido[1,2-a]pyrimidin-4-one, 9-Hydroxyrisperidone

P1897-1G P1897-200MG
TCIAP1897-1GEA 176 EUR
TCIAP1897-1G TCIAP1897-200MG
Paliperidone ≥97.0% (by titrimetric analysis)
Paliperidone
Formula: C₂₃H₂₇FN₄O₃
Peso molecolare: 426,49 g/mol
MDL Number: MFCD00871802
CAS Number: 144598-75-4
UN: 2811
ADR: 6.1,III

Visualizza i codici

Risultati del test di specificazione

Appearance Light orange to Yellow to Green powder to crystal
Purity (HPLC) min. 95.0 area%
Purity (Nonaqueous Titration) min. 97.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR