3-(1-Pyrrolylmethyl)pyridine ≥97.0% (by GC, titration analysis)
Fornitore: TCI
|
Attenzione
|
Formula:
C₁₀H₁₀N₂ |
MDL Number:
MFCD00006405 CAS Number: 80866-95-1 EINECS: 279-598-2 |
Risultati del test di specificazione
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 97.0 % |
Purity (Nonaqueous Titration) | min. 97.0 % |
Melting point | 58.0 to 62.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...