1,1'-(2,6-Pyridinediyl)bis(3-methylimidazolium)dibromide ≥98.0% (by HPLC, titration analysis)
Fornitore: TCI EUROPE
Synonyms:
1,1'-(Pyridine-2,6-diyl)bis(3-methyl-1H-imidazol-3-ium) bromide
P1816-1G
P1816-5G
TCIAP1816-1GEA
60
EUR
TCIAP1816-1G
TCIAP1816-5G
1,1'-(2,6-Pyridinediyl)bis(3-methylimidazolium)dibromide ≥98.0% (by HPLC, titration analysis)
1,1'-(2,6-Pyridinediyl)bis(3-methylimidazolium)dibromide
Formula:
C₁₃H₁₅Br₂N₅ Peso molecolare: 401,11 g/mol |
MDL Number:
MFCD11975436 CAS Number: 263874-05-1 |
Risultati del test di specificazione
Purity (HPLC) | min. 98.0 area% |
Purity (Nonaqueous Titration) | min. 98.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...